Learn More
ATCC FHC; Colon; Human (Homo sapiens)
Catalog No. 502384480
Encompass
FHC is an epithelial cell line that was isolated from the large intestine of a 13-week-old embryo. This cell line was deposited by DP Chopra.
Catalog No. 50-238-4480 Supplier ATCC Supplier No. CRL1831
FHC is an epithelial cell line that was isolated from the large intestine of a 13-week-old embryo. This cell line was deposited by DP Chopra.