Learn More
TARGETMOL CHEMICALS INC FHD-286 1MG
SDP
Catalog No. 502880373
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. FHD-286 is an inhibitor of BRM/BRG1 ATPase and can be used for studies on the treatment of BAF-related disorders such as acute myeloid leukemia. Purity 98.7%
Catalog No. 50-288-0373 Supplier TARGETMOL CHEMICALS INC Supplier No. T97491MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. FHD-286 is an inhibitor of BRM/BRG1 ATPase and can be used for studies on the treatment of BAF-related disorders such as acute myeloid leukemia. Purity 98.7%