Learn More
Medchemexpress LLC Fhd-609 | 2676211-64-4 | 99.2% | 829.00 | C47H56N8O6 | 10 MG
SDP
Catalog No. 5000287054
Encompass_Preferred
FHD-609 is a selective heterobifunctional degrader (PROTAC) of bromodomain-containing protein 9 (BRD9). It induces targeted proteasomal degradation via CRBN recruitment and is used in research to study BRD9 biology, ncBAF complex function, and oncology-related applications. The compound is supplied as a high-purity material for laboratory use and formulation work.
  • Selective BRD9 degrader and inhibitor.
  • High purity (>99%) suitable for research use.
  • Applicable to in vitro and preclinical studies.
  • Facilitates study of ncBAF complex and BRD9-related cancers.
  • Molecular weight 829.00 and formula C47H56N8O6.
Catalog No. 50-002-87054 Supplier Medchemexpress LLC Supplier No. HY15336710MG
FHD-609 is a selective heterobifunctional degrader (PROTAC) of bromodomain-containing protein 9 (BRD9). It induces targeted proteasomal degradation via CRBN recruitment and is used in research to study BRD9 biology, ncBAF complex function, and oncology-related applications. The compound is supplied as a high-purity material for laboratory use and formulation work.
  • Selective BRD9 degrader and inhibitor.
  • High purity (>99%) suitable for research use.
  • Applicable to in vitro and preclinical studies.
  • Facilitates study of ncBAF complex and BRD9-related cancers.
  • Molecular weight 829.00 and formula C47H56N8O6.