Learn More
Medchemexpress LLC FHD-609 | 2676211-64-4 | C47H56N8O6 | 500 MG
SDP
Catalog No. 5000372724
Encompass_Preferred
FHD-609 is a PROTAC degrader and inhibitor of BRD9 (Bromodomain-containing protein 9), targeting ncBAF. It can be utilized for research into various cancers featuring mutations in a BAF complex subunit. This compound has demonstrated sustained BRD9 degradation and anti-tumor activities in preclinical models. In combination with Telomelysin or INO5401, FHD-609 may contribute to the treatment of adrenocortical carcinoma (ACC).
  • Targets ncBAF
  • Used for research into cancers with BAF complex subunit mutations
  • Exhibits anti-tumor activities in preclinical models
  • May be useful in adrenocortical carcinoma (ACC) treatment when combined with other agents
Catalog No. 50-003-72724 Supplier Medchemexpress LLC Supplier No. HY153367500MG
FHD-609 is a PROTAC degrader and inhibitor of BRD9 (Bromodomain-containing protein 9), targeting ncBAF. It can be utilized for research into various cancers featuring mutations in a BAF complex subunit. This compound has demonstrated sustained BRD9 degradation and anti-tumor activities in preclinical models. In combination with Telomelysin or INO5401, FHD-609 may contribute to the treatment of adrenocortical carcinoma (ACC).
  • Targets ncBAF
  • Used for research into cancers with BAF complex subunit mutations
  • Exhibits anti-tumor activities in preclinical models
  • May be useful in adrenocortical carcinoma (ACC) treatment when combined with other agents