Learn More
Selleck Chemical LLC FHD-609-E1859-25MG
SDP
Catalog No. 503137761
Encompass_Preferred
FHD-609 acts as a selective inhibitor and degrader of BRD9 protein (bromodomain-containing protein 9) FHD-609 targets ncBAF and can be used in research of various cancers involving mutations in a BAF complex subunit
Catalog No. 50-313-7761 Supplier Selleck Chemical LLC Supplier No. E185925MG
FHD-609 acts as a selective inhibitor and degrader of BRD9 protein (bromodomain-containing protein 9) FHD-609 targets ncBAF and can be used in research of various cancers involving mutations in a BAF complex subunit