Learn More
Aviva Systems Biology Corp FHL1 Antibody - N-terminal region (ARP31902_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31902_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FHL1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Human. Alternate names: FHL1B, KYO-T, MGC111107, SLIM1, bA535K18.1, KYOT, SLIM, FHL-1, FHL1A, FLH1A, XMPMA, SLIM-1, SLIMMERApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-54361