Learn More
Aviva Systems Biology Corp FHL3 Antibody - middle region (ARP50498_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50498_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FHL3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FHL3. Alternate names: MGC19547, MGC23614, MGC8696, SLIM2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60358