Learn More
Aviva Systems Biology Corp FHL5 Antibody - middle region (ARP51673_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51673_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FHL5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FHL5. Alternate names: ACT, FLJ33049, KIAA0776, RP3-393D12.2, dJ393D12.2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60581