Learn More
ATCC FHM; Muscle; Fathead Minnow (Pimephales promelas)
Catalog No. 502382455
Encompass
FHM is an epithelial cell line that was isolated in 1962 from the connective tissue of an adult fathead minnow. This cell line was deposited by RG Malsberger.
Catalog No. 50-238-2455 Supplier ATCC Supplier No. CCL42
 Restricted Item
Add to Cart
Add to Cart
FHM is an epithelial cell line that was isolated in 1962 from the connective tissue of an adult fathead minnow. This cell line was deposited by RG Malsberger.