Learn More
Grainger FHP Sheave 2BK120 x 1-7/16
Catalog No. 502753224
Encompass_Preferred
The Grainger website features a wide selection of V-belt pulleys and idlers. These pulleys are used to transmit rotational mechanical power between two shafts using a V-belt. They are made from various materials like cast iron, steel, and aluminum. Pulleys come in different sizes, groove profiles, bores, and configurations like single, double, triple, and more. They also offer adjustable idlers to maintain proper belt tension. Overall, this page provides a comprehensive offering of high-quality V-belt pulleys and idlers for industrial power transmission applications.
Catalog No. 50-275-3224 Supplier Grainger Supplier No. 145P42
The Grainger website features a wide selection of V-belt pulleys and idlers. These pulleys are used to transmit rotational mechanical power between two shafts using a V-belt. They are made from various materials like cast iron, steel, and aluminum. Pulleys come in different sizes, groove profiles, bores, and configurations like single, double, triple, and more. They also offer adjustable idlers to maintain proper belt tension. Overall, this page provides a comprehensive offering of high-quality V-belt pulleys and idlers for industrial power transmission applications.