Learn More
Grainger FHP Sheave 2BK72-QT
Catalog No. 502749045
Encompass_Preferred
The V-Belt Sheave Bushed Bore from Baldor is a V-belt pulley with a 1-1/8" bore and a 3/4" keyway. It has an outside diameter of 4-1/2" and is made of cast iron with a rough bore finish. This sheave is designed for use with v-belts and has a bushing installed in the bore to reduce wear. The maximum bore RPM is 3600. This is a high quality, durable V-belt sheave well-suited for industrial applications requiring a fixed bore pulley.
Catalog No. 50-274-9045 Supplier Grainger Supplier No. 145P77
The V-Belt Sheave Bushed Bore from Baldor is a V-belt pulley with a 1-1/8" bore and a 3/4" keyway. It has an outside diameter of 4-1/2" and is made of cast iron with a rough bore finish. This sheave is designed for use with v-belts and has a bushing installed in the bore to reduce wear. The maximum bore RPM is 3600. This is a high quality, durable V-belt sheave well-suited for industrial applications requiring a fixed bore pulley.