Learn More
Grainger FHP Sheave AK24 x 1
Catalog No. 502752319
Encompass_Preferred
The Approved Vendor V-Belt Pulley is a 3.4" diameter x 1.4" face width pulley made of cast iron with a set screw. It has a 0.5" bore and is used with v-belts. This pulley can be utilized for power transmission applications and features a black finish. The overall dimensions are 3.4" diameter x 1.4" face width x 1.25" hub projection. It is rated for up to 1/2 horsepower at 1750 RPM. The pulley's set screw and keyway help lock it into position on a shaft.
Catalog No. 50-275-2319 Supplier Grainger Supplier No. 145R32
The Approved Vendor V-Belt Pulley is a 3.4" diameter x 1.4" face width pulley made of cast iron with a set screw. It has a 0.5" bore and is used with v-belts. This pulley can be utilized for power transmission applications and features a black finish. The overall dimensions are 3.4" diameter x 1.4" face width x 1.25" hub projection. It is rated for up to 1/2 horsepower at 1750 RPM. The pulley's set screw and keyway help lock it into position on a shaft.