Learn More
Grainger FHP Sheave AK30 x 1
Catalog No. 502752876
Encompass_Preferred
The Sheave Fixed Bore 145R47 is a sheave with a fixed bore size of 1-7/16 inches. It is made of cast iron with a black oxide finish. The sheave has a 4-1/2 inch outside diameter and is 1 inch wide. It has a load capacity of 2750 pounds with a maximum RPM of 1725. The sheave requires a 1-7/16 inch shaft and has set screws for mounting. It weighs approximately 5 pounds and is manufactured in the USA by Baldor. The sheave is commonly used for power transmission applications.
Catalog No. 50-275-2876 Supplier Grainger Supplier No. 145R47
The Sheave Fixed Bore 145R47 is a sheave with a fixed bore size of 1-7/16 inches. It is made of cast iron with a black oxide finish. The sheave has a 4-1/2 inch outside diameter and is 1 inch wide. It has a load capacity of 2750 pounds with a maximum RPM of 1725. The sheave requires a 1-7/16 inch shaft and has set screws for mounting. It weighs approximately 5 pounds and is manufactured in the USA by Baldor. The sheave is commonly used for power transmission applications.