Learn More
Grainger FHP Sheave AK35 x 7/8
Catalog No. 502750556
Encompass_Preferred
The Approved Vendor V-Belt Pulley is a 3L V-belt pulley with a 1-7/16" bore size and 1/2" keyway. It is constructed of cast iron with a black oxide finish. This pulley has an OD of 4-1/2", is 1-3/4" wide, and works with v-belts sized AX, BX, and CX. It can be used for power transmission applications and features set screws for securing to a shaft. Made in the USA.
Catalog No. 50-275-0556 Supplier Grainger Supplier No. 145R59
The Approved Vendor V-Belt Pulley is a 3L V-belt pulley with a 1-7/16" bore size and 1/2" keyway. It is constructed of cast iron with a black oxide finish. This pulley has an OD of 4-1/2", is 1-3/4" wide, and works with v-belts sized AX, BX, and CX. It can be used for power transmission applications and features set screws for securing to a shaft. Made in the USA.