Learn More
Grainger FHP Sheave AK84-QT
Catalog No. 502753563
Encompass_Preferred
The V-Belt Sheave Bushed Bore 145T20 is a V-belt pulley with a bushing bore. It has a 2.25 inch outside diameter, a 0.75 inch bore diameter, and a 0.25 inch bushing length. The sheave is made of cast iron with a rough finish and operates at speeds up to 6580 feet per minute. It is designed for use with fractional horsepower drives. The bushing provides support for the shaft and allows for adjustment of the sheave on the shaft. This sheave enables power transmission in industrial applications via a v-belt drive system.
Catalog No. 50-275-3563 Supplier Grainger Supplier No. 145T20
The V-Belt Sheave Bushed Bore 145T20 is a V-belt pulley with a bushing bore. It has a 2.25 inch outside diameter, a 0.75 inch bore diameter, and a 0.25 inch bushing length. The sheave is made of cast iron with a rough finish and operates at speeds up to 6580 feet per minute. It is designed for use with fractional horsepower drives. The bushing provides support for the shaft and allows for adjustment of the sheave on the shaft. This sheave enables power transmission in industrial applications via a v-belt drive system.