Learn More
Grainger FHP Sheave BK47 x 3/4
Catalog No. 502749237
Encompass_Preferred
The Approved Vendor V-Belt Pulley is a 3L V-belt pulley with a 1-7/8" bore size and 1/2" keyway. It is made from cast iron with a black oxide finish and has a maximum speed rating of 3600 RPM. This pulley can be used with v-belts sized A, B, C, and D. It features set screws for securing to a shaft. The overall dimensions are 3-1/2" diameter and 2-3/4" width. This heavy duty pulley provides a rigid drive for industrial power transmission applications.
Catalog No. 50-274-9237 Supplier Grainger Supplier No. 145U15
The Approved Vendor V-Belt Pulley is a 3L V-belt pulley with a 1-7/8" bore size and 1/2" keyway. It is made from cast iron with a black oxide finish and has a maximum speed rating of 3600 RPM. This pulley can be used with v-belts sized A, B, C, and D. It features set screws for securing to a shaft. The overall dimensions are 3-1/2" diameter and 2-3/4" width. This heavy duty pulley provides a rigid drive for industrial power transmission applications.