Learn More
ATCC FHs 74 Int; Small Intestine; Human (Homo sapiens)
Catalog No. 502383080
Encompass
FHs 74 Int is a cell line exhibiting epithelial morphology isolated from the small intestine of a 3 to 4 months gestation, female patient. This cell line was deposited by R Owens.
Catalog No. 50-238-3080 Supplier ATCC Supplier No. CCL241
FHs 74 Int is a cell line exhibiting epithelial morphology isolated from the small intestine of a 3 to 4 months gestation, female patient. This cell line was deposited by R Owens.