Learn More
Medchemexpress LLC FHT-1015 | 2368903-18-6 | 98.4% | C25H25N5O4S3 | 50 MG
SDP
Catalog No. 5000354121
Encompass_Preferred
FHT-1015 is a selective inhibitor of SMARCA4 and SMARCA2, also known as BRG1 and BRM. It functions as an allosteric inhibitor, inducing a conformational change in the BRG1/BRM protein at an allosteric site, thereby inhibiting ATPase activity. This compound has been shown to interfere with tumor cell growth and migration and is a subject of research for uveal melanoma and hematologic cancer.
  • Interferes with tumor cell growth and migration
  • Subject of research for uveal melanoma
  • Subject of research for hematologic cancer
Catalog No. 50-003-54121 Supplier Medchemexpress LLC Supplier No. HY14489650MG
FHT-1015 is a selective inhibitor of SMARCA4 and SMARCA2, also known as BRG1 and BRM. It functions as an allosteric inhibitor, inducing a conformational change in the BRG1/BRM protein at an allosteric site, thereby inhibiting ATPase activity. This compound has been shown to interfere with tumor cell growth and migration and is a subject of research for uveal melanoma and hematologic cancer.
  • Interferes with tumor cell growth and migration
  • Subject of research for uveal melanoma
  • Subject of research for hematologic cancer