Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC FHT-2344 | 2468058-90-2 | 98.5% | 528.60 | 100 MG
SDP

Catalog No. 5000360839
Encompass_Preferred

FHT-2344 is a chemical probe and a SMARCA4/SMARCA2 ATPase inhibitor, demonstrating anticancer activity. It has IC50 values of 0.026 μM and 0.013 μM for SMARCA4 and SMARCA2 ATPase, respectively.

  • High purity
  • Chemical formula: C23H24N6O5S2
  • Appearance: solid, white to off-white
  • Soluble in DMSO (100 mg/mL)
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • Solution storage: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-60839 Supplier Medchemexpress LLC Supplier No. HY149458100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FHT-2344 is a chemical probe and a SMARCA4/SMARCA2 ATPase inhibitor, demonstrating anticancer activity. It has IC50 values of 0.026 μM and 0.013 μM for SMARCA4 and SMARCA2 ATPase, respectively.

  • High purity
  • Chemical formula: C23H24N6O5S2
  • Appearance: solid, white to off-white
  • Soluble in DMSO (100 mg/mL)
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • Solution storage: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.