Learn More
Medchemexpress LLC Fht-2344 | 2468058-90-2 | 98.5% | 528.60 | 5 MG
SDP
Catalog No. 5000360240
Encompass_Preferred
FHT-2344 is a chemical probe that acts as a SMARCA4/SMARCA2 ATPase inhibitor. It exhibits anticancer activity.
  • Acts as a SMARCA4/SMARCA2 ATPase inhibitor with IC50 values of 0.026 μM and 0.013 μM
  • Exhibits anticancer activity
Catalog No. 50-003-60240 Supplier Medchemexpress LLC Supplier No. HY1494585MG
FHT-2344 is a chemical probe that acts as a SMARCA4/SMARCA2 ATPase inhibitor. It exhibits anticancer activity.
  • Acts as a SMARCA4/SMARCA2 ATPase inhibitor with IC50 values of 0.026 μM and 0.013 μM
  • Exhibits anticancer activity