Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC FHT-2344 | 2468058-90-2 | 98.5% | 528.60 | 50 MG
SDP

Catalog No. 5000360836
Encompass_Preferred

FHT-2344 is a chemical probe and a SMARCA4/SMARCA2 ATPase inhibitor with IC50 values of 0.026 μM and 0.013 μM, respectively. It also exhibits anticancer activity. It is for research use only.

  • SMARCA4/SMARCA2 ATPase inhibitor
  • Anticancer activity
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-60836 Supplier Medchemexpress LLC Supplier No. HY14945850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FHT-2344 is a chemical probe and a SMARCA4/SMARCA2 ATPase inhibitor with IC50 values of 0.026 μM and 0.013 μM, respectively. It also exhibits anticancer activity. It is for research use only.

  • SMARCA4/SMARCA2 ATPase inhibitor
  • Anticancer activity
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.