Learn More
Innovative Research Inc FI Fbrngn Plsmngn Dplt Pur 1gm
Supplier:  Innovative Research Inc IHUFBGPLGD1GM
Encompass_Preferred
Human Factor I (Fibrinogen) Plasminogen Depleted Purified 1gm - It is purified from human plasma and plasminogen depleted using affinity chromatography. This is a frozen liquid buffered in 20 mM sodium citrate-HCl/ pH 7.4. The Human Fibrinogen is homogeneous as observed on a 4-20% gradient gel, and is greater than 95% clottable. Fibrinogen (FI) naturally circulates in the blood and is a key factor in clotting and coagulation. Fibrinogen is converted to Fibrin by Thrombin, which leads to the development of a fibrin-based clot. Fibrinogen also promotes clotting by bridging and activating platelets. As a positive acute-phase protein, Fibrinogen levels will rise in response to inflammation, injury, and other events, and can be elevated in the presence of some cancers. Purity Homogeneous as observed on a 4-20% gradient gel, > 95% clottable Storage Conditions -60 to -80C. Thaw in a 37C water bath, then aliquot and freeze the unused portion at -60C or below.
Catalog No. 50-274-4338