Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp WATERS CORPORATION Standards, FI optimization, GC, MS, 1 mL

Catalog No. 50466517 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This Routine Use MS Standard is specifically for GCTs and designed for FI optimization on GCTs only. It contains Perfluorotrimethylcyclohexane Heptacosa Hexafluorobenzene Pentafluorobenzene Chloropentafluorobenzene Xylene and Acetone.

  • Separation mode: GC
  • Detection method: MS
  • Formulation: liquid
  • Storage condition: room temperature
  • System type: GCMS
  • Shelf life: 12 months
  • Shipping condition: ambient
  • Volume: 1 mL
  • Type: calibration
  • Product type: standards
  • Units per package: 1 pk
  • Contains hazardous material which requires special freight handling, and additional charges may apply.

Catalog No. 50-466-517 Supplier Waters Corp Supplier No. 186006865
May include imposed supplier surcharges.
Add to Cart
Add to Cart

This Routine Use MS Standard is specifically for GCTs and designed for FI optimization on GCTs only. It contains Perfluorotrimethylcyclohexane Heptacosa Hexafluorobenzene Pentafluorobenzene Chloropentafluorobenzene Xylene and Acetone.

  • Separation mode: GC
  • Detection method: MS
  • Formulation: liquid
  • Storage condition: room temperature
  • System type: GCMS
  • Shelf life: 12 months
  • Shipping condition: ambient
  • Volume: 1 mL
  • Type: calibration
  • Product type: standards
  • Units per package: 1 pk
  • Contains hazardous material which requires special freight handling, and additional charges may apply.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.