Learn More
LabStrong Corp FI-STREEM 8 LITER CARBOY
Catalog No. 50551560
Encompass_Preferred
water purification/distillation system accessories, Fi-Streem Distillation 8 liter storage carboy,Compatible with LabStrong FI-Streem II 2S glass still; 17D x 8W x 6.5 in. H (43 x 20 x 17cm); Produced by LabStong Corporation. Model A1052LS
Catalog No. 50551560 Supplier LabStrong Corp Supplier No. A1052LS
water purification/distillation system accessories, Fi-Streem Distillation 8 liter storage carboy,Compatible with LabStrong FI-Streem II 2S glass still; 17D x 8W x 6.5 in. H (43 x 20 x 17cm); Produced by LabStong Corporation. Model A1052LS