Learn More
LabStrong Corp FI-STREEM DRAIN/FILL SOLENOID
Catalog No. NC0308511
Encompass_Preferred
LabStrong Fi-Streem part number 857X22LS compares to Fi-Streem part number 857X22. The solenoid valve (drain/fill, 24V AC) is a replacement part for Fi-Streem Water Purification Stills.
Catalog No. NC0308511 Supplier LabStrong Corp Supplier No. 857X22LS
LabStrong Fi-Streem part number 857X22LS compares to Fi-Streem part number 857X22. The solenoid valve (drain/fill, 24V AC) is a replacement part for Fi-Streem Water Purification Stills.