Learn More
LabStrong Corp FI-STREEM GLASS BOILER
Catalog No. NC0020977
Encompass_Preferred
LabStrong part number 562X2LS compares to Fi-Streem part number 562X2. The glass boiler is a replacement part for Fi-Streem Water Purification Stills. The glass boiler comes with one 562X77LS (Silicone Sleeve, Large, 50mm length) and one 562X64LS (Silicone Sleeve, Small, 35mm length).
Catalog No. NC0020977 Supplier LabStrong Corp Supplier No. 562X2LS
LabStrong part number 562X2LS compares to Fi-Streem part number 562X2. The glass boiler is a replacement part for Fi-Streem Water Purification Stills. The glass boiler comes with one 562X77LS (Silicone Sleeve, Large, 50mm length) and one 562X64LS (Silicone Sleeve, Small, 35mm length).