Learn More
LabStrong Corp FI-STREEM HEAT ELEMENT 562X23
Catalog No. NC0010662
Encompass_Preferred
The heating element (240V, 1.5 kW) is a replacement part for Fi-Streem Water Purification Stills. The heating element includes the plastic element union and heater sheath.
Catalog No. NC0010662 Supplier LabStrong Corp Supplier No. 562X23LS
The heating element (240V, 1.5 kW) is a replacement part for Fi-Streem Water Purification Stills. The heating element includes the plastic element union and heater sheath.