Learn More
LabStrong Corp FI-STREEM HEATING ELEMENT 240V
Catalog No. NC0039632
Encompass_Preferred
The LabStrong part number EL744X4LS compares to Fi-Streem part number EL744X4. The heating element (220V, 900W) is a replacement part for Fi-Streem Water Purification Stills. The heating element includes the plastic element union and heater sheath.
Catalog No. NC0039632 Supplier LabStrong Corp Supplier No. EL744X4LS
The LabStrong part number EL744X4LS compares to Fi-Streem part number EL744X4. The heating element (220V, 900W) is a replacement part for Fi-Streem Water Purification Stills. The heating element includes the plastic element union and heater sheath.