Learn More
LabStrong Corp FI-STREEM II STILL 4 L/HR 220V
Catalog No. NC1058161
Encompass_Preferred
Of all the water purification technologies available, distillation can remove a wider range of impurities than any other single form of water purification. LabStrong Fi-Streem Stills consistently produce high quality ASTM Type II water and can remove inorganic ions, dissolved gases, organics, particles, bacteria and pyrogens from water. LabStrong Fi-Streem Water Stills will meet your laboratory water needs while remaining budget-friendly. The LabStrong Fi-Streem 2S Still consistently produces high quality ASTM Type II water. The still produces four liters of distilled water per hour and comes with a one year limited warranty. Product Features: Consistently Pure Water The still consistently produces high quality ASTM Type II water and can remove inorganic ions, dissolved gases, organics, particles, bacteria and pyrogens from water.
Catalog No. NC1058161 Supplier LabStrong Corp Supplier No. A74420LS
Add to Cart
Add to Cart
Request a Quote
Of all the water purification technologies available, distillation can remove a wider range of impurities than any other single form of water purification. LabStrong Fi-Streem Stills consistently produce high quality ASTM Type II water and can remove inorganic ions, dissolved gases, organics, particles, bacteria and pyrogens from water. LabStrong Fi-Streem Water Stills will meet your laboratory water needs while remaining budget-friendly. The LabStrong Fi-Streem 2S Still consistently produces high quality ASTM Type II water. The still produces four liters of distilled water per hour and comes with a one year limited warranty. Product Features: Consistently Pure Water The still consistently produces high quality ASTM Type II water and can remove inorganic ions, dissolved gases, organics, particles, bacteria and pyrogens from water.