Learn More
OCEAN OPTICS INC FIA FLOW CELLS
Catalog No. NC2242576
Encompass
Stainless steel. Internal Volume: Up to 26 µLPathlength: Adjustable up to 10 mmWavelength Range: 210nm - 2μm
Catalog No. NC2242576 Supplier OCEAN OPTICS INC Supplier No. FIA1000ZSS
Stainless steel. Internal Volume: Up to 26 µLPathlength: Adjustable up to 10 mmWavelength Range: 210nm - 2μm