Learn More
Selleck Chemical LLC Fialuridine-E1709-5MG
SDP
Catalog No. 503138815
Encompass_Preferred
Fialuridine(FIAU DRG-0098 NSC 678514) is a nucleoside analog with antiviral activity It blocks DNAsynthesis in human cytomegalovirus and hepatitis B as well as herpes simplex
Catalog No. 50-313-8815 Supplier Selleck Chemical LLC Supplier No. E17095MG
Fialuridine(FIAU DRG-0098 NSC 678514) is a nucleoside analog with antiviral activity It blocks DNAsynthesis in human cytomegalovirus and hepatitis B as well as herpes simplex