Learn More
Mitegen FIB Autogrid Rings With High-Contrast Orientation Marks By Nanosoft
SDP
Catalog No. 5000495017
Encompass
Nanosoft FIB Autogrid Rings with high-contrast orientation marks. A significant increase in the area of your grid is now accessible for milling with the ion beam, as the Nanosoft ring features an angled face around its entire circumference. This allows you to reclaim valuable sample and cells on your grid that would otherwise be inaccessible. Additionally, the rings have high-contrast markings on both the bottom and top faces, making correct orientation incredibly easy. This eliminates the need for manual marking and ensures precise alignment. Manufactured with cutting-edge techniques and individually checked by experts, these rings provide top-quality, consistent, and reliable parts, sold in packs of 100.
Catalog No. 50-004-95017 Supplier Mitegen Supplier No. MNSFIBARC01
Nanosoft FIB Autogrid Rings with high-contrast orientation marks. A significant increase in the area of your grid is now accessible for milling with the ion beam, as the Nanosoft ring features an angled face around its entire circumference. This allows you to reclaim valuable sample and cells on your grid that would otherwise be inaccessible. Additionally, the rings have high-contrast markings on both the bottom and top faces, making correct orientation incredibly easy. This eliminates the need for manual marking and ensures precise alignment. Manufactured with cutting-edge techniques and individually checked by experts, these rings provide top-quality, consistent, and reliable parts, sold in packs of 100.