Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Fiber 20 Gal Lock Rim, Poly Cover w/GSKT
SDP

Catalog No. 19823212
Change view
Click to view available options
Capacity (English):
20 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19-823-212 20 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-823-212 Supplier Youngstown Barrel & Drum Supplier No. 1995P
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fiber 20 Gal Lock Rim, Poly Cover w/GSKT. MOQ of 100/ea. along with at least minimums of like drums listed, totaling a truckload quantity. Part numbers 191305584, 191305585, 191305586, 19171191. UN rated 1G/Y105/S. 15 in. dia x 25-1/4 in.H.

Specifications

Product Type Fiber Drum
Closure Type Lever Lock Ring
Description New Fiber 20 Gal Lock Rim, Poly Cover w/GSKT UN1G/Y105/S; Fiber / Ppr 275
Capacity (English) 20 gal.
Material Fiber
Head Style Open
Height (English) 25.2362205 in.
Height (Metric) 641 mm
Diameter (English) Inner 15.6692913 in.
Diameter (English) Outer 16.1417323 in.
Diameter (Metric) Inner 398 mm
Diameter (Metric) Outer 410mm
Weight (English) 7.51776315 lb.
Weight (Metric) 75.7082356 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.