Learn More
Grainger Fiber Cord Duplex SC SC 1m Orange
Catalog No. 502752415
Encompass_Preferred
The Grainger fiber optic cable patch cords are high-speed, high-bandwidth cables for transmitting data, voice, and video over long distances. They use light signals instead of electrical signals, enabling faster speeds and longer distances without signal loss. These cables have LC connectors on each end and are available in singlemode and multimode with various fiber counts. Lengths range from 1 to 30 meters. Key features include low attenuation, electromagnetic interference resistance, and compatibility with 10G networks. These durable, flexible cables are ideal for data centers, telecom rooms, network cabling projects and more.
Catalog No. 50-275-2415 Supplier Grainger Supplier No. 14C133
The Grainger fiber optic cable patch cords are high-speed, high-bandwidth cables for transmitting data, voice, and video over long distances. They use light signals instead of electrical signals, enabling faster speeds and longer distances without signal loss. These cables have LC connectors on each end and are available in singlemode and multimode with various fiber counts. Lengths range from 1 to 30 meters. Key features include low attenuation, electromagnetic interference resistance, and compatibility with 10G networks. These durable, flexible cables are ideal for data centers, telecom rooms, network cabling projects and more.