Learn More
Grainger Fiber Cord Duplex ST ST 2m Orange
Catalog No. 502752419
Encompass_Preferred
The Grainger fiber optic cable patch cords are available in single-mode and multimode options with SC, ST, LC, and MTRJ connectors. They feature OFNP plenum-rated jackets and are designed for high-speed data transmission, supporting 10G Ethernet, Fibre Channel, ATM, or SONET networks. The cables have low loss and support long transmission distances. Lengths range from 1 to 50 feet. The patch cords are durable, flexible, and RoHS compliant. They are ideal for connecting network equipment in data centers, telecom closets, and work areas.
Catalog No. 50-275-2419 Supplier Grainger Supplier No. 14C124
The Grainger fiber optic cable patch cords are available in single-mode and multimode options with SC, ST, LC, and MTRJ connectors. They feature OFNP plenum-rated jackets and are designed for high-speed data transmission, supporting 10G Ethernet, Fibre Channel, ATM, or SONET networks. The cables have low loss and support long transmission distances. Lengths range from 1 to 50 feet. The patch cords are durable, flexible, and RoHS compliant. They are ideal for connecting network equipment in data centers, telecom closets, and work areas.