Learn More
Grainger ARC ABRASIVES Fiber Disc, Predator, Ceramic, 120 Grit, 25/Pk.
Catalog No. 502773085
Encompass_Preferred
These fiber discs are designed for effective grinding and finishing applications on aluminum and non-ferrous materials. Engineered for durability and high-performance, they feature a ceramic abrasive grain and a robust fiber backing to ensure consistent results during demanding tasks.
  • Abrasive material: ceramic
  • Abrasive grade: 120 grit (medium)
  • Coating type: closed
  • Arbor size: 7/8 in.
  • Maximum operating speed: 8,600 RPM
  • Resistance properties: heat and load resistant
  • Vacuum hole design: no hole
  • Package quantity: 25
Catalog No. 50-277-3085 Supplier Grainger Supplier No. 13T169
These fiber discs are designed for effective grinding and finishing applications on aluminum and non-ferrous materials. Engineered for durability and high-performance, they feature a ceramic abrasive grain and a robust fiber backing to ensure consistent results during demanding tasks.
  • Abrasive material: ceramic
  • Abrasive grade: 120 grit (medium)
  • Coating type: closed
  • Arbor size: 7/8 in.
  • Maximum operating speed: 8,600 RPM
  • Resistance properties: heat and load resistant
  • Vacuum hole design: no hole
  • Package quantity: 25