Learn More
MSC World Centric Food Container Lids, Square, Unbleached Plant Fiber, 400/Pack
Catalog No. 502815706
Encompass
These food container lids are designed for use with World Centric TRSC60 containers. Made from unbleached plant fiber, they are square-shaped and feature a snap-on design for secure closure.
  • Compostable in two to four months in a commercial composting facility
  • Made from bamboo and unbleached plant fiber, an annually renewable resource
  • No added PFAS
  • USDA Certified Biobased 100%
  • BPI Certified Compostable
Catalog No. 50-281-5706 Supplier MSC Supplier No. 14171987
These food container lids are designed for use with World Centric TRSC60 containers. Made from unbleached plant fiber, they are square-shaped and feature a snap-on design for secure closure.
  • Compostable in two to four months in a commercial composting facility
  • Made from bamboo and unbleached plant fiber, an annually renewable resource
  • No added PFAS
  • USDA Certified Biobased 100%
  • BPI Certified Compostable