Learn More
Poltex Fiber Optic Laryngoscope Countertop Blade Holder
SDP
Catalog No. 22000333
Change view
Click to view available options
Dimensions (L x W x H):
10.5 x 9 x 5 in.
Catalog No. Dimensions (L x W x H)
22-000-333 10.5 x 9 x 5 in.
1 options
Catalog No. 22-000-333 Supplier Poltex Supplier No. LRYNG4CT
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart
Works in conjunction with Poltex Fiber Optic Laryngoscope Blade Holster
  • Holds blade handle and back up blades for fiber optic laryngoscope
  • Durable PETG construction
  • Non-slip rubber bumpons on base
  • Easily cleanable
  • Alcohol resistant
Specifications
Autoclavable Not Autoclavable
Holds Holds blade handle and back up blades for fiber optic laryngoscope
For Use With (Application) Works in conjunction with HS-18116
Mounting Countertop
Dimensions (L x W x H) 10.5 x 9 x 5 in.