Learn More
Surgical Design™ Fiber Optic Laryngoscope Handles and Blades (Mac and Miller)
Shop All Surgical Design ProductsDescription
- Complete range of fiber optic laryngoscope handles: Large, Medium, and Small
- Available Macintosh fiber optic blades (005) and Miller fiber optic blades (005)
- Integrated fiber optic light transmission provides bright, cool, shadow-free illumination
- High-quality stainless steel with polished optical window for maximum light output
- Ideal for airway visualization, difficult airway assessment, emergency response, and postmortem airway inspection
- Anti-corrosive, easy-clean design suitable for repeated autoclave sterilization
- Secure, stable blade-to-handle locking system
- Knurled ergonomic handle ensures reliable grip in clinical and morgue environments
- Compatible with most standard fiber optic systems and medical lighting setups
- Smooth blade contours minimize tissue trauma during use
Specifications
Specifications
| Product Type | Laryngoscope Blade, Miller |
| Material | Fiber Optic |
| Sterility | Non-sterile |
| Disposable | Reusable |
| Size | #5 |