Learn More
Kent Scientific Fiber Optic Lighting
SDP
Catalog No. 13005169 Shop All Kent Scientific Products
Encompass
Change view
Click to view available options
Animal:
Mouse
Catalog No. Animal
13-005-169 Mouse
1 options
Catalog No. 13-005-169 Supplier Kent Scientific Supplier No. ETIMSE02
Includes
25 in. fiber optic cable, trans-tracheal illuminator, LED light, endotracheal tubes/wedges, USB cable, A/C adapter
For use with mouse intubation kit.
  • For use with mouse intubation kit
Specifications
Animal Mouse
For Use With (Application) Mouse Intubation Kit
Type Fiber Optic Lighting
Includes 25 in. fiber optic cable, trans-tracheal illuminator, LED light, endotracheal tubes/wedges, USB cable, A/C adapter