Learn More
World Precision Instrument Fiber Optic Oxygen Meter for Microsensors
SDP
Catalog No. 502538640
Encompass
A generation of sensors based on luminescence lifetime
The measurement principle of the sensor system is based on the detection of oxygen concentration as a function of luminescence lifetime, either in dissolved or gaseous phase environments.
Catalog No. 50-253-8640 Supplier World Precision Instrument Supplier No. OXYMICROAOT
Add to Cart
Add to Cart
Request a Quote
A generation of sensors based on luminescence lifetime
The measurement principle of the sensor system is based on the detection of oxygen concentration as a function of luminescence lifetime, either in dissolved or gaseous phase environments.