Learn More
Electron Microscopy Sciences Ultratool Fiber Polisher KIT
SDP
Catalog No. 501926741
Encompass
Designed for manual polishing of fibers, either supported (connectorized) or nonsupported (bare fibers), the tool uses various replaceable modules which are available for all types of connectors. Key to ULTRATOOL's performance is a height adjust feature that is controlled by a fine pitch micrometer screw thread. Flat, PC & APC polishes can be performed on all standard connector types. Another common use is bare-fiber polishing applications (flat and angled). ULTRATOOL is capable of holding bare fibers of the full size range and many other materials such as small wafer sections (for planar or edge polishes). Tool, wear stops, connector inserts, locking tool, ULTRAPOL B.
Catalog No. 50-192-6741 Supplier Electron Microscopy Sciences Supplier No. 1101.6
Designed for manual polishing of fibers, either supported (connectorized) or nonsupported (bare fibers), the tool uses various replaceable modules which are available for all types of connectors. Key to ULTRATOOL's performance is a height adjust feature that is controlled by a fine pitch micrometer screw thread. Flat, PC & APC polishes can be performed on all standard connector types. Another common use is bare-fiber polishing applications (flat and angled). ULTRATOOL is capable of holding bare fibers of the full size range and many other materials such as small wafer sections (for planar or edge polishes). Tool, wear stops, connector inserts, locking tool, ULTRAPOL B.