Learn More
Hach Company Fiber Wiper TU5 Auto Clean Mod
Supplier:  Hach Company LZQ176
Encompass
Built to handle samples containing iron and manganese the fiber wiper for the TU5300sc and TU5400sc Automatic Cleaning Module maintains a clean sample cell for your turbidity measurements
Catalog No. 50-002-07065