Learn More
Andwin Scientific FIBERBOARD DIVIDERS 5X5X1
SDP
Catalog No. NC9370141
Encompass_Preferred
The CryoPro Fiberboard cardboard cell divider feature numbered grids that allow users to have easy identification and organization of items. This product measures 12.3 x 12.3 x 3.2 cm (417/20 x 417/20 x 11/4 “), with each cell measuring 1.3 cm (0.53"). There are 81 cells in this product. It is made of durable fiberboard with a protective, moisture-repellant coating. This product is sold in a pack of 12.
Catalog No. NC9370141 Supplier Andwin Scientific Supplier No. 82021126
The CryoPro Fiberboard cardboard cell divider feature numbered grids that allow users to have easy identification and organization of items. This product measures 12.3 x 12.3 x 3.2 cm (417/20 x 417/20 x 11/4 “), with each cell measuring 1.3 cm (0.53"). There are 81 cells in this product. It is made of durable fiberboard with a protective, moisture-repellant coating. This product is sold in a pack of 12.