Learn More
Mopec Fiberglass Latent Print Powder Brush
SDP
Catalog No. 22446335 Shop All Mopec Products
Change view
Click to view available options
For Use With (Application):
Latent Fingerprint Development
Catalog No. For Use With (Application)
22-446-335 Latent Fingerprint Development
1 options
Catalog No. 22-446-335 Supplier Mopec Supplier No. FS348
Add to Cart
Add to Cart
Soft fiberglass bristles apply fingerprint powder for developing latent prints while allowing controlled, even handling. A balanced plastic handle supports comfortable application, and an anti-static storage tube protects the bristles and keeps the tool contained between uses.
Construction
  • Print powder brush
  • Anti-static tube
Dimensions
  • Handle: 4.13 in. L
  • Bristles: 2.25 in. L
  • Tube: 7.5 in. L
Materials
  • Fiberglass
  • Plastic
Specifications
Product Type Latent Print Brush
For Use With (Application) Latent Fingerprint Development