Learn More
MSC Made In USA Air Filter Media Pad, Fiberglass, 20 in. x 20 in. x 2 in., 50/Pk.
Catalog No. 5000507869
Encompass
These fiberglass air filter media pads are designed for depth loading, efficiently capturing dust, dirt, pollen, and other foreign particles in air handling units, furnaces, and air intake systems. The media features a progressively dense structure, starting with a loosely woven front layer for larger debris and concluding with a dense back layer for fine particulate removal.
  • Fiberglass media with a light oil coating to enhance contaminant collection
  • Progressively dense structure for depth loading and high dust holding capacity
  • Designed for use as a primary pre-filter or as a final filter
  • Initial resistance of 0.22 in. w.g. at 350 FPM
  • MERV 7 efficiency rating with 84% arrestance
  • Maximum operating temperature of 150°F
Catalog No. 50-005-07869 Supplier MSC Supplier No. 59828004
These fiberglass air filter media pads are designed for depth loading, efficiently capturing dust, dirt, pollen, and other foreign particles in air handling units, furnaces, and air intake systems. The media features a progressively dense structure, starting with a loosely woven front layer for larger debris and concluding with a dense back layer for fine particulate removal.
  • Fiberglass media with a light oil coating to enhance contaminant collection
  • Progressively dense structure for depth loading and high dust holding capacity
  • Designed for use as a primary pre-filter or as a final filter
  • Initial resistance of 0.22 in. w.g. at 350 FPM
  • MERV 7 efficiency rating with 84% arrestance
  • Maximum operating temperature of 150°F