Learn More
Brady™ Fiberglass Sign: CAUTION
Catalog No. 19104741 Shop All Brady Corporation Products
Change view
Click to view available options
Material:
Fiberglass
Catalog No. Material
19-104-741 Fiberglass
1 options
Catalog No. 19-104-741 Supplier Brady™ Supplier No. 76005
  • $51.25
    $30.65 / Each of 1
    Save $20.60
    40% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Improve workplace safety with proper signage and markings. This Brady™ fiberglass sign is a durable, clear and effective tool to raise awareness to workplace hazards.
  • Fiberglass material is abrasion-resistant and chemical-resistant
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • Fiberglass out-performs all other sign materials in terms of strength, durability, and longevity
  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use
Specifications
Material Fiberglass
Mounting Type Corner Holes
Shape Rectangular
Languages English
Dimensions (L x W) 10 x 7 in.
Displayed Text CAUTION
Product Type Safety Sign
Color Black on Yellow
WARNING: Cancer - www.P65Warnings.ca.gov