Learn More
MSC MFG Tray Storage Tote, Stack and Nest, Fiberglass, 300 lb Capacity, 1 ea
Catalog No. 5000510944
Encompass
This stack and nest storage tote is constructed from high-strength fiberglass reinforced composite, designed to provide exceptional durability and support for heavy loads in industrial environments. Featuring a 300 lb weight capacity, this container allows for efficient space management by stacking when full and nesting when empty. Its robust material composition resists bending, sagging, denting, and corrosion, making it suitable for demanding applications.
  • Constructed from high-strength fiberglass reinforced composite
  • Features a stack and nest design for space efficiency
  • Supports a weight capacity of 300 lb
  • Resists cutting oils, detergents, mild acids, and alkaline solutions
  • Operates in a temperature range of -60 °F to 250 °F
  • Designed with smooth surfaces and radial edges for safe handling
  • Ideal for machine integration and robotic applications
Catalog No. 50-005-10944 Supplier MSC Supplier No. 79042545
This stack and nest storage tote is constructed from high-strength fiberglass reinforced composite, designed to provide exceptional durability and support for heavy loads in industrial environments. Featuring a 300 lb weight capacity, this container allows for efficient space management by stacking when full and nesting when empty. Its robust material composition resists bending, sagging, denting, and corrosion, making it suitable for demanding applications.
  • Constructed from high-strength fiberglass reinforced composite
  • Features a stack and nest design for space efficiency
  • Supports a weight capacity of 300 lb
  • Resists cutting oils, detergents, mild acids, and alkaline solutions
  • Operates in a temperature range of -60 °F to 250 °F
  • Designed with smooth surfaces and radial edges for safe handling
  • Ideal for machine integration and robotic applications