Learn More
Grainger Fiberglass Stair Tread 144 x 12 In
Catalog No. 502766456
Encompass_Preferred
The Fibergrate Fiberglass Bar Stair Tread is a heavy duty, corrosion resistant stair tread made of molded fiberglass reinforced plastic with a meniscus bar surface. It has a width of 24 inches, length of 36 inches, thickness of 1.5 inches, and weight of 11 lbs. The tread features a slip resistant surface and is designed for industrial and commercial use on metal pan stairs. It meets OSHA and ADA requirements. The bar surface provides maximum slip resistance.
Catalog No. 50-276-6456 Supplier Grainger Supplier No. 16U525
The Fibergrate Fiberglass Bar Stair Tread is a heavy duty, corrosion resistant stair tread made of molded fiberglass reinforced plastic with a meniscus bar surface. It has a width of 24 inches, length of 36 inches, thickness of 1.5 inches, and weight of 11 lbs. The tread features a slip resistant surface and is designed for industrial and commercial use on metal pan stairs. It meets OSHA and ADA requirements. The bar surface provides maximum slip resistance.