Learn More
Grainger Fiberglass Stair Tread 36 x 12 In
Catalog No. 502764877
Encompass_Preferred
The Fibergrate Fiberglass Bar Stair Tread is a heavy duty stair tread made of molded fiberglass reinforced plastic with a meniscus bar surface. It has a slip resistant surface and is designed for industrial and commercial use on stairs and landings. This stair tread measures 36 inches long x 11-1/2 inches wide x 1-3/8 inches thick and weighs 9 lbs. It can support a uniform load of 100 psf. The bar spacing is 1-3/16 inches on center. This stair tread comes in safety yellow color and has a flame spread rating less than 25. It is manufactured in the USA by Fibergrate.
Catalog No. 50-276-4877 Supplier Grainger Supplier No. 16U533
The Fibergrate Fiberglass Bar Stair Tread is a heavy duty stair tread made of molded fiberglass reinforced plastic with a meniscus bar surface. It has a slip resistant surface and is designed for industrial and commercial use on stairs and landings. This stair tread measures 36 inches long x 11-1/2 inches wide x 1-3/8 inches thick and weighs 9 lbs. It can support a uniform load of 100 psf. The bar spacing is 1-3/16 inches on center. This stair tread comes in safety yellow color and has a flame spread rating less than 25. It is manufactured in the USA by Fibergrate.